Learn More
ILEX LIFE SCIENCES LLC PAN-Biotech DMEM/F12 (1:1) Cell Culture Medium, w: L-Glutamine, w: 15 mM HEPES, w: 1.2 g/L NaHCO3 (500 ml)
SDP
Catalog No. 502388524
Encompass_Preferred
PAN-Biotech DMEM/F12 (1:1) Cell Culture Medium, w: L-Glutamine, w: 15 mM HEPES, w: 1.2 g/L NaHCO3. QUANTITY: 500 ml. MANUFACTURED BY: PAN-Biotech GmbH (Germany). BICARBONATE: With 1.2 g/L Sodium Bicarbonate. GLUCOSE: with glucose. GLUTAMINE: with L-Glutamine. HEPES: With 15 mM HEPES. PHENOL RED: With Phenol Red. SODIUM PYRUVATE: with Sodium pyruvate. STERILE: Yes. STORAGE TEMPERATURE: +2-8°C. SUMMARY: DMEM/F12 Medium is a 1:1 mixture of Dulbecco’s Modified Essential Medium (DMEM) and Ham’s F-12 Medium, commonly used in growing a variety of mammalian cell cultures. DMEM/F12 is rich and complex, containing all 21 amino acids, 10 vitamins, glucose, zinc, and iron among other components. Together, it combines DMEM's high concentration of amino acids and vitamins of DMEM and the higher concentration of zinc sulfate, putrescine and linoleic acid of F12.
Catalog No. 50-238-8524 Supplier ILEX LIFE SCIENCES LLC Supplier No. P0441250
PAN-Biotech DMEM/F12 (1:1) Cell Culture Medium, w: L-Glutamine, w: 15 mM HEPES, w: 1.2 g/L NaHCO3. QUANTITY: 500 ml. MANUFACTURED BY: PAN-Biotech GmbH (Germany). BICARBONATE: With 1.2 g/L Sodium Bicarbonate. GLUCOSE: with glucose. GLUTAMINE: with L-Glutamine. HEPES: With 15 mM HEPES. PHENOL RED: With Phenol Red. SODIUM PYRUVATE: with Sodium pyruvate. STERILE: Yes. STORAGE TEMPERATURE: +2-8°C. SUMMARY: DMEM/F12 Medium is a 1:1 mixture of Dulbecco’s Modified Essential Medium (DMEM) and Ham’s F-12 Medium, commonly used in growing a variety of mammalian cell cultures. DMEM/F12 is rich and complex, containing all 21 amino acids, 10 vitamins, glucose, zinc, and iron among other components. Together, it combines DMEM's high concentration of amino acids and vitamins of DMEM and the higher concentration of zinc sulfate, putrescine and linoleic acid of F12.