Learn More
ATHENA ES SILAC- DMEM High Glucose, powder
Catalog No. NC2235015
Encompass_Preferred
SILAC media is prepared specifically for labeling experiments involving the use of stable amino acid isotopes. Athena provides several of the most commonly used media without L-arginine, L-lysine, L-leucine and L-methionine, the four mostly commonly used heavy isotope labeled amino acids.
This item is not returnable. View return policy
Catalog No. NC2235015 Supplier ATHENA ES Supplier No. 0430
SILAC media is prepared specifically for labeling experiments involving the use of stable amino acid isotopes. Athena provides several of the most commonly used media without L-arginine, L-lysine, L-leucine and L-methionine, the four mostly commonly used heavy isotope labeled amino acids.