Learn More
Fisherbrand™ DMEM/High Glucose, with 4.0 mM L-Glutamine, 4500 mg/L Glucose, and Sodium Pyruvate
Shop All Fisherbrand ProductsDescription
DMEM (Dulbecco's Modified Eagle Medium) is a widely used medium supporting many mammalian cell lines including but not limited to HeLa and 293 cells.
This DMEM is modified with:
- 4500 mg/L of glucose
- 4.0 mM L-Glutamine
- Sodium pyruvate
Specifications
Specifications
| Cell Line | Primary Fibroblasts, Neurons, Glial Cells, HUVECs, Smooth Muscle Cells |
| Cell Type | HeLa, 293, Cos-7, and PC-12 |
| Classification | None |
| Form | Liquid |
| Product Type | DMEM (Dulbecco's Modified Eagle Medium) |
| Serum Level | Standard Serum Supplementation |
| Sterility | Sterile |
| Sterilization Method | 0.1 μm Sterile-filtered |
| With Additives | Glutamine, Glucose, Sodium Pyruvate |
| Without Additives | None |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.