Learn More
ILEX LIFE SCIENCES LLC DMEM P04-01501 6 X 500 ML
SDP
Catalog No. 502733895
Encompass_Preferred
PAN-Biotech DMEM Cell Culture Medium, w: 1.0 g/L Glucose, w/o: L-Glutamine, w: Sodium pyruvate, w/o: Calcium, w: 3.7 g/L NaHCO3 (6 x 500 ml). BICARBONATE: With 3.7 g/L Sodium Bicarbonate. GLUCOSE: Low Glucose (1.0 g/L). GLUTAMINE: No glutamine. HEPES: Without HEPES. PHENOL RED: With Phenol Red. SODIUM PYRUVATE: With Sodium Pyruvate. CALCIUM: Without Calcium. STERILE: Yes. STORAGE TEMP.: +2-8°C. MANUFACTURER: PAN-Biotech GmbH. COUNTRY OF ORIGIN: Germany. DESCRIPTION: Intrinsically developed for the cultivation of murine embryonic cells, DMEM (Dulbecco's Modified Eagle Medium) is tailor-made for the cultivation of a broad range of cells, especially if the medium is supplemented with FBS. DMEM is an Eagle medium modification with a fourfold content of amino acids and vitamins. DMEM with 1.0 g/L Glucose is the standard medium, whereas DMEM with 4.5 g/L Glucose is for cells which have a high energy demand.
Catalog No. 50-273-3895 Supplier ILEX LIFE SCIENCES LLC Supplier No. P0401501X6
Add to Cart
Add to Cart
PAN-Biotech DMEM Cell Culture Medium, w: 1.0 g/L Glucose, w/o: L-Glutamine, w: Sodium pyruvate, w/o: Calcium, w: 3.7 g/L NaHCO3 (6 x 500 ml). BICARBONATE: With 3.7 g/L Sodium Bicarbonate. GLUCOSE: Low Glucose (1.0 g/L). GLUTAMINE: No glutamine. HEPES: Without HEPES. PHENOL RED: With Phenol Red. SODIUM PYRUVATE: With Sodium Pyruvate. CALCIUM: Without Calcium. STERILE: Yes. STORAGE TEMP.: +2-8°C. MANUFACTURER: PAN-Biotech GmbH. COUNTRY OF ORIGIN: Germany. DESCRIPTION: Intrinsically developed for the cultivation of murine embryonic cells, DMEM (Dulbecco's Modified Eagle Medium) is tailor-made for the cultivation of a broad range of cells, especially if the medium is supplemented with FBS. DMEM is an Eagle medium modification with a fourfold content of amino acids and vitamins. DMEM with 1.0 g/L Glucose is the standard medium, whereas DMEM with 4.5 g/L Glucose is for cells which have a high energy demand.