Learn More
ILEX LIFE SCIENCES LLC DMEM P04-05540 6 X 500 ML
SDP
Catalog No. 502733871
Encompass_Preferred
PAN-Biotech DMEM Cell Culture Medium, w: 4.5 g/L Glucose, w: L-Glutamine, w: 25 mM HEPES, w/o: Sodium pyruvate, w: 3.7 g/L NaHCO3 (6 x 500 ml). BICARBONATE: With 3.7 g/L Sodium Bicarbonate. GLUCOSE: High glucose (4.5 g/L). GLUTAMINE: With L-glutamine. HEPES: With 25mM HEPES. PHENOL RED: With Phenol Red. SODIUM PYRUVATE: Without Sodium Pyruvate. STERILE: Yes. STORAGE TEMP.: +2-8°C. MANUFACTURER: PAN-Biotech GmbH. COUNTRY OF ORIGIN: Germany. DESCRIPTION: Intrinsically developed for the cultivation of murine embryonic cells, DMEM (Dulbecco's Modified Eagle Medium) is tailor-made for the cultivation of a broad range of cells, especially if the medium is supplemented with FBS. DMEM is an Eagle medium modification with a fourfold content of amino acids and vitamins. DMEM with 1.0 g/L Glucose is the standard medium, whereas DMEM with 4.5 g/L Glucose is for cells which have a high energy demand.
Catalog No. 50-273-3871 Supplier ILEX LIFE SCIENCES LLC Supplier No. P0405540X6
Add to Cart
Add to Cart
PAN-Biotech DMEM Cell Culture Medium, w: 4.5 g/L Glucose, w: L-Glutamine, w: 25 mM HEPES, w/o: Sodium pyruvate, w: 3.7 g/L NaHCO3 (6 x 500 ml). BICARBONATE: With 3.7 g/L Sodium Bicarbonate. GLUCOSE: High glucose (4.5 g/L). GLUTAMINE: With L-glutamine. HEPES: With 25mM HEPES. PHENOL RED: With Phenol Red. SODIUM PYRUVATE: Without Sodium Pyruvate. STERILE: Yes. STORAGE TEMP.: +2-8°C. MANUFACTURER: PAN-Biotech GmbH. COUNTRY OF ORIGIN: Germany. DESCRIPTION: Intrinsically developed for the cultivation of murine embryonic cells, DMEM (Dulbecco's Modified Eagle Medium) is tailor-made for the cultivation of a broad range of cells, especially if the medium is supplemented with FBS. DMEM is an Eagle medium modification with a fourfold content of amino acids and vitamins. DMEM with 1.0 g/L Glucose is the standard medium, whereas DMEM with 4.5 g/L Glucose is for cells which have a high energy demand.