Learn More
Quality Biological Inc DMEM with Glutamine XL™, Case 10 X 500 mL
SDP
Catalog No. 502209743
Encompass_Preferred
DMEM (Dulbeccos Modified Eagle Medium) is a widely used basal medium for supporting the growth of many different mammalian cells. Cells successfully cultured in DMEM include primary fibroblasts neurons glial cells HUVECs and smooth muscle cells. We offer a variety of DMEM modifications for a range of cell culture applications.
With stable L-alanyl-glutamine (Glutamine XL)
Manufactured under an ISO 9001:2015 certified quality management system.
Product and/or packaging contains recycled/renewable content. Product and/or packaging is recyclable or reduces waste. Product and/or packaging is safer to human and environmental health.
Catalog No. 50-220-9743 Supplier Quality Biological Inc Supplier No. 112300101CS
DMEM (Dulbeccos Modified Eagle Medium) is a widely used basal medium for supporting the growth of many different mammalian cells. Cells successfully cultured in DMEM include primary fibroblasts neurons glial cells HUVECs and smooth muscle cells. We offer a variety of DMEM modifications for a range of cell culture applications.
With stable L-alanyl-glutamine (Glutamine XL)
Manufactured under an ISO 9001:2015 certified quality management system.
Product and/or packaging contains recycled/renewable content. Product and/or packaging is recyclable or reduces waste. Product and/or packaging is safer to human and environmental health.