Learn More
Sigma Aldrich Fine Chemicals Biosciences Dulbecco's Modified Eagle's Medium Without glucose, L-glutamine, phenol red, sodium pyruvate and sodium bicarbonate, powder, suitable for cell culture | 10X1L
Catalog No. NC1671539
Encompass
Dulbecco's Modified Eagle's Medium Without glucose, L-glutamine, phenol red, sodium pyruvate and sodium bicarbonate, powder, suitable for cell culture | 10X1L
Catalog No. NC1671539 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. D503010X1L
Dulbecco's Modified Eagle's Medium Without glucose, L-glutamine, phenol red, sodium pyruvate and sodium bicarbonate, powder, suitable for cell culture | 10X1L