Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ DMEM, low glucose, pyruvate
GREENER_CHOICE

Catalog No. 11885076
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 mL
1000 mL
10 x 500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-885-076 1000 mL
11-885-084 500 mL
11-885-092 10 x 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 11-885-076 Supplier Gibco™ Supplier No. 11885076
Only null left
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy

Requires

Supplementation

DMEM (Dulbecco's Modified Eagle Medium) is a widely used basal medium for supporting the growth of many different mammalian cells.

DMEM (Dulbecco's Modified Eagle Medium) is a widely used basal medium for supporting the growth of many different mammalian cells. Cells successfully cultured in DMEM include primary fibroblasts, neurons, glial cells, HUVECs, and smooth muscle cells, as well as cell lines such as HeLa, 293, Cos-7, and PC-12. We offer a variety of DMEM modifications for a range of cell culture applications. Find the right formulation using the media selector tool.

This DMEM is modified as follows:

With: Low Glucose, L-Glutamine, Sodium Pyruvate, Phenol Red

Without: HEPES

The complete formulation is available.

Using DMEM

DMEM is unique from other media as it contains 4 times the concentration of amino acids and vitamins than the original Eagle's Minimal Essential Medium. DMEM was originally formulated with low glucose (1 g/L) and sodium pyruvate, but is often used with higher glucose levels, with or without sodium pyruvate. DMEM contains no proteins, lipids, or growth factors. Therefore, DMEM requires supplementation, commonly with 10% Fetal Bovine Serum (FBS). DMEM uses a sodium bicarbonate buffer system (3.7 g/L), and therefore requires a 5–10% CO2 environment to maintain physiological pH.

Product use

For human ex vivo tissue and cell culture processing applications. CAUTION: When used as a medical device, Federal law restricts this device to sale by or on the order of a physician. Customers using Gibco™ DMEM in a manufacturing process, who have a submission with the FDA, may request a letter of authorization from us to reference our Type II Drug Master File (DMF).

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Cell Line HeLa, 293, Cos-7, and PC-12
Cell Type Primary Fibroblasts, Neurons, Glial Cells, HUVECs, Smooth Muscle Cells
Classification Animal Origin-free
Concentration 1 X
Form Liquid
Product Type DMEM (Dulbecco's Modified Eagle Medium)
Serum Level Standard Serum Supplementation
Sterility Sterile-filtered
Sterilization Method Sterile-filtered
With Additives Low Glucose, Glutamine, Phenol Red, Sodium Pyruvate
Without Additives No HEPES
Manufacturing Quality cGMP-compliant under the ISO 13485 standard
Product Line Gibco
Quantity 1000 mL
Shipping Condition Room Temperature
Content And Storage Storage conditions: 2-8° C. Protect from light
Shipping conditions: Ambient
Shelf life: 12 months from date of manufacture
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.