Learn More
Enzo Life Sciences DMEQ-TAD (100mg). CAS: 132788-52-4
SDP
Catalog No. 502013704
Encompass_Preferred
DMEQ-TAD is a reagent for fluorescence-labeling of Vitamin D and its use in assaying Vitamin D metabolites. Alternative name: 4-[2-(6,7-Dimethoxy-4-methyl-3-oxo-3,4-dihydroquinoxalinyl)ethyl]-1,2,4-triazoline-3,5-dione. Purity: ≥98% (TLC). Solubility: Soluble in dichloromethane, acetone or chloroform. Long Term Storage: -20°C.
Catalog No. 50-201-3704 Supplier Enzo Life Sciences Supplier No. ENZCHM2490100
DMEQ-TAD is a reagent for fluorescence-labeling of Vitamin D and its use in assaying Vitamin D metabolites. Alternative name: 4-[2-(6,7-Dimethoxy-4-methyl-3-oxo-3,4-dihydroquinoxalinyl)ethyl]-1,2,4-triazoline-3,5-dione. Purity: ≥98% (TLC). Solubility: Soluble in dichloromethane, acetone or chloroform. Long Term Storage: -20°C.