Learn More
Sigma Aldrich Fine Chemicals Biosciences Dmethyl sulfoxde Reagent500ML
Catalog No. 502899944
Encompass
Dimethyl sulfoxide is an amphipathic molecule soluble in both aqueous and organic solvents. It is commonly used as an aprotic solvent and also as a reagent in organic synthesis.
Catalog No. 50-289-9944 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. D5879500ML
Dimethyl sulfoxide is an amphipathic molecule soluble in both aqueous and organic solvents. It is commonly used as an aprotic solvent and also as a reagent in organic synthesis.