Learn More
Selleck Chemical LLC DMF (Dimethyl Fumarate) S2586-1g
SDP
Catalog No. 503112545
Encompass_Preferred
DMF (Dimethyl Fumarate) is the methyl ester of fumaric acid used to treat people with relapsing forms of multiple sclerosis Dimethyl fumarate is a nuclear factor (erythroid-derived)-like 2 (Nrf2) pathway activator and induces upregulation of antioxidant gene expression
Catalog No. 50-311-2545 Supplier Selleck Chemical LLC Supplier No. S25861G
DMF (Dimethyl Fumarate) is the methyl ester of fumaric acid used to treat people with relapsing forms of multiple sclerosis Dimethyl fumarate is a nuclear factor (erythroid-derived)-like 2 (Nrf2) pathway activator and induces upregulation of antioxidant gene expression