Learn More
Selleck Chemical LLC DMF
SDP
Catalog No. 5000000079
Encompass_Preferred
DMF(N N-Dimethylformamide) is a polar organic solvent with a wide range of industrial applications This product is a hazardous chemical (acute toxicity/flammable/skin corrosive) Please use it while wearing a protective face mask gloves and clothing
Catalog No. 50-000-00079 Supplier Selleck Chemical LLC Supplier No. S61925ML
DMF(N N-Dimethylformamide) is a polar organic solvent with a wide range of industrial applications This product is a hazardous chemical (acute toxicity/flammable/skin corrosive) Please use it while wearing a protective face mask gloves and clothing