Learn More
Medchemexpress LLC Dmf-dg 500mg
SDP
Catalog No. 5000206199
Encompass_Preferred
Dmf-dg (2 -Deoxy-N2-dimethylaminomethylene-guanosine) is a deoxyguanosine (dG) nucleoside protected by the dimethylaminomethylamidine (DMF) base and can be used for oligonucleotide synthesis[1]
Catalog No. 50-002-06199 Supplier Medchemexpress LLC Supplier No. HYW098689500MG
Dmf-dg (2 -Deoxy-N2-dimethylaminomethylene-guanosine) is a deoxyguanosine (dG) nucleoside protected by the dimethylaminomethylamidine (DMF) base and can be used for oligonucleotide synthesis[1]