Learn More
Description
- DMF for sequencing and peptide synthesis
- Useful for protein chemistry due to low amine content
- DMF is a commonly used solvent in solid phase peptide synthesis
- Intended for research use only
Specifications
Specifications
| pH | 6 to 8 |
| Melting Point | -61°C |
| Absorbance | Pass Test, 1 max. at 270nm, 0.3 max. at 275nm, 0.1 max. at 295nm, 0.05 max. at 310nm, 0.01 max. (340nm to 400nm) at 340nm |
| Color | Colorless |
| Decomposition Information | >350°C |
| Density | 0.945g/cm³ |
| Quantity | 500 mL |
| Identification | Pass Test |
| Packaging | Glass Bottle |
| Physical Form | Liquid |
| Show More |
Safety and Handling
| Hazard Statement | WARNING! |
| Precautionary Statement | Emergency Overview Flammable liquid and vapor. May cause methemoglobinemia. Irritating to eyes and skin. Lachrymator (substance which increases the flow of tears). May cause central nervous system effects. May cause adverse liver effects. May cause adverse kidney effects. May cause harm to the unborn child. Use personal protective equipment. Keep away from open flames |
| DOTInformation | DOT Class 3, : Flammable Liquid |
| Recommended Storage | RT |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.