Learn More
Medchemexpress LLC Dmg-peg 2000 | 160743-62-4 | 98.4% | 2000 (Average) | 100 MG
SDP
Catalog No. 5000437781
Encompass_Preferred
DMG-PEG 2000 is used for the preparation of liposomes for siRNA delivery, improving transfection efficiency in vitro. It is also utilized in lipid nanoparticles for an oral plasmid DNA delivery approach in vivo through surface modification, which enhances the mucus permeability and delivery efficiency of the nanoparticles. The product is for research use only and not sold to patients.
  • Used for liposome preparation for siRNA delivery with improved in vitro transfection efficiency.
  • Used for lipid nanoparticles in oral plasmid DNA delivery, improving mucus permeability and delivery efficiency in vivo.
Catalog No. 50-004-37781 Supplier Medchemexpress LLC Supplier No. HY112764100MG
DMG-PEG 2000 is used for the preparation of liposomes for siRNA delivery, improving transfection efficiency in vitro. It is also utilized in lipid nanoparticles for an oral plasmid DNA delivery approach in vivo through surface modification, which enhances the mucus permeability and delivery efficiency of the nanoparticles. The product is for research use only and not sold to patients.
  • Used for liposome preparation for siRNA delivery with improved in vitro transfection efficiency.
  • Used for lipid nanoparticles in oral plasmid DNA delivery, improving mucus permeability and delivery efficiency in vivo.