Learn More
Medchemexpress LLC DMG-PEG 2000 | 160743-62-4 | 98.4% | 2000 (Average) | 25 MG
SDP
Catalog No. 5000439346
Encompass_Preferred
DMG-PEG 2000 is utilized for the preparation of liposomes for siRNA delivery, leading to improved transfection efficiency in vitro. Additionally, it is employed in lipid nanoparticles for an oral plasmid DNA delivery approach in vivo. This involves a facile surface modification to enhance the mucus permeability and delivery efficiency of the nanoparticles.
  • Used for the preparation of liposome for siRNA delivery with improved transfection efficiency in vitro.
  • Used for lipid nanoparticle for an oral plasmid DNA delivery approach in vivo through facile surface modification.
  • Improves mucus permeability and delivery efficiency of nanoparticles.
Catalog No. 50-004-39346 Supplier Medchemexpress LLC Supplier No. 25MGHY112764
DMG-PEG 2000 is utilized for the preparation of liposomes for siRNA delivery, leading to improved transfection efficiency in vitro. Additionally, it is employed in lipid nanoparticles for an oral plasmid DNA delivery approach in vivo. This involves a facile surface modification to enhance the mucus permeability and delivery efficiency of the nanoparticles.
  • Used for the preparation of liposome for siRNA delivery with improved transfection efficiency in vitro.
  • Used for lipid nanoparticle for an oral plasmid DNA delivery approach in vivo through facile surface modification.
  • Improves mucus permeability and delivery efficiency of nanoparticles.