Learn More
Medchemexpress LLC Dmg-peg 2000 | 160743-62-4 | 98.4% | 2000 (Average) | 50 MG
SDP
Catalog No. 5000437369
Encompass_Preferred
DMG-PEG 2000 is utilized in the preparation of liposomes for siRNA delivery, enhancing transfection efficiency in vitro. It is also employed for lipid nanoparticles in an oral plasmid DNA delivery approach in vivo, through surface modification, to improve mucus permeability and delivery efficiency of the nanoparticles.
  • Used for liposome preparation for siRNA delivery with improved transfection efficiency in vitro.
  • Used for lipid nanoparticles for oral plasmid DNA delivery with improved mucus permeability and delivery efficiency in vivo.
Catalog No. 50-004-37369 Supplier Medchemexpress LLC Supplier No. HY11276450MG
DMG-PEG 2000 is utilized in the preparation of liposomes for siRNA delivery, enhancing transfection efficiency in vitro. It is also employed for lipid nanoparticles in an oral plasmid DNA delivery approach in vivo, through surface modification, to improve mucus permeability and delivery efficiency of the nanoparticles.
  • Used for liposome preparation for siRNA delivery with improved transfection efficiency in vitro.
  • Used for lipid nanoparticles for oral plasmid DNA delivery with improved mucus permeability and delivery efficiency in vivo.