Learn More
Avanti Polar Lipids DMG-PEG 2000, 160743-62-4, powder, 1g
Catalog No. NC1665314
Encompass
Molecular Formula C122H242O50 (average MW due to polydispersity of PEG), Purity >99%, Formula Weight 2509.200, Storage Temperature -20, 1,2-dimyristoyl-rac-glycero-3-methoxypolyethylene glycol-2000
Catalog No. NC1665314 Supplier Avanti Polar Lipids Supplier No. 880151P1G
Molecular Formula C122H242O50 (average MW due to polydispersity of PEG), Purity >99%, Formula Weight 2509.200, Storage Temperature -20, 1,2-dimyristoyl-rac-glycero-3-methoxypolyethylene glycol-2000