Learn More
TARGETMOL CHEMICALS INC DMG-PEG 2000 200MG
SDP
Catalog No. 502579021
Encompass_Preferred
Also available in 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. DMG-PEG 2000 (DMG-PEG2000) is a lipid excipient that has been used in combination with other lipids in the formation of lipid nanoparticles. Formulations containing DMG-PEG 2000 have been used in the development of lipid nanoparticles for the delivery of mRNA-based vaccines. purity: 98%
Catalog No. 50-257-9021 Supplier TARGETMOL CHEMICALS INC Supplier No. T9098200MG
Also available in 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. DMG-PEG 2000 (DMG-PEG2000) is a lipid excipient that has been used in combination with other lipids in the formation of lipid nanoparticles. Formulations containing DMG-PEG 2000 have been used in the development of lipid nanoparticles for the delivery of mRNA-based vaccines. purity: 98%