Learn More
Medchemexpress LLC HY-112764 500mg Medchemexpress, DMG-PEG 2000 CAS:160743-62-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY112764500MG
Encompass_Preferred
Medchemexpress, HY-112764 500mg DMG-PEG 2000 CAS:160743-62-4 DMG-PEG 2000 is used for the preparation of liposome for siRNA delivery with improved transfection efficiency in vitro. DMG-PEG 2000 is also used for the lipid nanoparticle for an oral plasmid DNA delivery approach in vivo through a facile surface modification to improve the mucus permeability and delivery efficiency of the nanoparticles. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9741