Learn More
Avanti Polar Lipids DMG-PEG(2k)-DBCO, Powder, 25 mg, C140H254N2O51, >99% Purity, Stability 1 year, Not Hygroscopic, Not Light Sensitive
Catalog No. NC3233141
Encompass
DMG-PEG(2k)-DBCO is a specialized polyethylene glycol (PEG) lipid designed for advanced lipid nanoparticle (LNP) applications. This clickable PEG lipid facilitates the conjugation of specific cell-targeting functional groups to LNPs via click chemistry, enabling targeted payload delivery to desired cells. The ability to vary PEG length with similar compounds influences micelle size, immunological safety, and drug release rates.
  • Designed for advanced lipid nanoparticle applications
  • Facilitates conjugation of cell-targeting groups
  • Enables targeted payload delivery
  • Varied PEG length influences micelle size and drug release
  • High purity of over 99%
  • Stable for 1 year
  • Not hygroscopic
  • Not light sensitive
Catalog No. NC3233141 Supplier Avanti Polar Lipids Supplier No. 880242P25MG
DMG-PEG(2k)-DBCO is a specialized polyethylene glycol (PEG) lipid designed for advanced lipid nanoparticle (LNP) applications. This clickable PEG lipid facilitates the conjugation of specific cell-targeting functional groups to LNPs via click chemistry, enabling targeted payload delivery to desired cells. The ability to vary PEG length with similar compounds influences micelle size, immunological safety, and drug release rates.
  • Designed for advanced lipid nanoparticle applications
  • Facilitates conjugation of cell-targeting groups
  • Enables targeted payload delivery
  • Varied PEG length influences micelle size and drug release
  • High purity of over 99%
  • Stable for 1 year
  • Not hygroscopic
  • Not light sensitive