Learn More
Apexbio Technology LLC DMG-PEG-MAL-MW-2000-25MG
SDP
Catalog No. 5000567827
Encompass_Preferred
DMG-PEG-Mal (MW 2000) is an essential reagent in biomedical research commonly used in drug delivery systems and bio-conjugation studies It originates from the conjugation of dimyristoyl glycerol (DMG) polyethylene glycol (PEG) and maleimide (Mal) enabling targeted delivery by linking therapeutic agents to specific biomolecules Its mechanism focuses on thiol-reactive maleimide bonds facilitating precise modification of proteins and peptides Known for its in vitro activity in the nanomolar to low micromolar range it is widely applied in cell models animal studies and drug screening Experimental concentrations typically depend on the specific research design
Catalog No. 50-005-67827 Supplier Apexbio Technology LLC Supplier No. M201425
DMG-PEG-Mal (MW 2000) is an essential reagent in biomedical research commonly used in drug delivery systems and bio-conjugation studies It originates from the conjugation of dimyristoyl glycerol (DMG) polyethylene glycol (PEG) and maleimide (Mal) enabling targeted delivery by linking therapeutic agents to specific biomolecules Its mechanism focuses on thiol-reactive maleimide bonds facilitating precise modification of proteins and peptides Known for its in vitro activity in the nanomolar to low micromolar range it is widely applied in cell models animal studies and drug screening Experimental concentrations typically depend on the specific research design