Learn More
Medchemexpress LLC DMG-PEG-Mal | 99.3% | 2000 (Average) | 10 MG
SDP
Catalog No. 5000289017
Encompass_Preferred
DMG-PEG-Mal (MW 2000) is a maleimide-functionalized PEGylated lipid used to improve delivery efficiency and tissue specificity of poorly soluble drugs. It is commonly used in formulation research such as lipid nanoparticles and liposomes, where the maleimide group enables conjugation to thiol-containing ligands.
  • Molecular weight 2000 (average).
  • Maleimide-functionalized PEG backbone for thiol conjugation.
  • High purity (about 99.3%).
  • White to off-white solid appearance.
  • Available in small research quantities including 10 mg.
  • Powder storage: -20°C (stable for up to 3 years); in solvent: -80°C for 6 months or -20°C for 1 month.
  • Intended for research use only; not for human use.
Catalog No. 50-002-89017 Supplier Medchemexpress LLC Supplier No. HY16703610MG
DMG-PEG-Mal (MW 2000) is a maleimide-functionalized PEGylated lipid used to improve delivery efficiency and tissue specificity of poorly soluble drugs. It is commonly used in formulation research such as lipid nanoparticles and liposomes, where the maleimide group enables conjugation to thiol-containing ligands.
  • Molecular weight 2000 (average).
  • Maleimide-functionalized PEG backbone for thiol conjugation.
  • High purity (about 99.3%).
  • White to off-white solid appearance.
  • Available in small research quantities including 10 mg.
  • Powder storage: -20°C (stable for up to 3 years); in solvent: -80°C for 6 months or -20°C for 1 month.
  • Intended for research use only; not for human use.