Learn More
Medchemexpress LLC Dmg-peg-nhs | 95.0% | 2000 (Average) | 25 MG
SDP
Catalog No. 5000384019
Encompass_Preferred
This compound is composed of Myristic acid and PEG linked by an amide bond, with NHS as an active ester group attached to the end of the PEG chain. It is suitable for various applications including protein/biomolecule modification, gene delivery, and biosensor applications.
  • Used for protein/biomolecule modification
  • Suitable for gene delivery
  • Applicable in biosensor applications
  • For research use only
Catalog No. 50-003-84019 Supplier Medchemexpress LLC Supplier No. 25MGHY174264
This compound is composed of Myristic acid and PEG linked by an amide bond, with NHS as an active ester group attached to the end of the PEG chain. It is suitable for various applications including protein/biomolecule modification, gene delivery, and biosensor applications.
  • Used for protein/biomolecule modification
  • Suitable for gene delivery
  • Applicable in biosensor applications
  • For research use only