Learn More
Medchemexpress LLC Dmg-PEG-NHS | 95.0% | 2000 Da | 10 MG
SDP
Catalog No. 5000298507
Encompass_Preferred
NHS-activated polyethylene glycol (PEG) linker terminated with a dimyristoyl glycerol (DMG) lipid moiety and an N-hydroxysuccinimide (NHS) ester. Average molecular weight ~2,000 Da. Supplied as a white to off-white solid for research use; the NHS ester enables amine-reactive conjugation to proteins and other biomolecules and supports liposome and gene-delivery formulations.
  • Enables efficient amine-reactive conjugation to proteins and peptides.
  • Suitable for liposome formulation and surface PEGylation.
  • Facilitates gene delivery and nucleic acid encapsulation.
  • White to off-white solid, easy to handle and weigh.
  • Stored as powder at -20°C for long-term stability; in-solution storage limited.
  • High purity by NMR (95.0%), appropriate for research applications.
Catalog No. 50-002-98507 Supplier Medchemexpress LLC Supplier No. HY17426410MG
NHS-activated polyethylene glycol (PEG) linker terminated with a dimyristoyl glycerol (DMG) lipid moiety and an N-hydroxysuccinimide (NHS) ester. Average molecular weight ~2,000 Da. Supplied as a white to off-white solid for research use; the NHS ester enables amine-reactive conjugation to proteins and other biomolecules and supports liposome and gene-delivery formulations.
  • Enables efficient amine-reactive conjugation to proteins and peptides.
  • Suitable for liposome formulation and surface PEGylation.
  • Facilitates gene delivery and nucleic acid encapsulation.
  • White to off-white solid, easy to handle and weigh.
  • Stored as powder at -20°C for long-term stability; in-solution storage limited.
  • High purity by NMR (95.0%), appropriate for research applications.