Learn More
Medchemexpress LLC DMG-PEG-NHS, MW 2000 | 95.0% | 2000 | 100 MG
SDP
Catalog No. 5000386193
Encompass_Preferred
DMG-PEG-NHS, MW 2000 is a compound composed of Myristic acid (HY-N2041) and PEG linked by an amide bond, with NHS as an active ester group attached to the end of the PEG chain. It is for research use only.
  • Used for protein/biomolecule modification
  • Suitable for gene delivery applications
  • Applicable in biosensor development
Catalog No. 50-003-86193 Supplier Medchemexpress LLC Supplier No. HY174264100MG
DMG-PEG-NHS, MW 2000 is a compound composed of Myristic acid (HY-N2041) and PEG linked by an amide bond, with NHS as an active ester group attached to the end of the PEG chain. It is for research use only.
  • Used for protein/biomolecule modification
  • Suitable for gene delivery applications
  • Applicable in biosensor development