Learn More
Medchemexpress LLC DMG-PEG-NHS, MW 2000 | 95.0% | 2000 | 5 MG
SDP
Catalog No. 5000386729
Encompass_Preferred
DMG-PEG-NHS, MW 2000 is a compound composed of Myristic acid (HY-N2041) and PEG linked by an amide bond, with NHS as an active ester group attached to the end of the PEG chain.
  • Used for protein/biomolecule modification.
  • Supports gene delivery.
  • Applicable in biosensor applications.
Catalog No. 50-003-86729 Supplier Medchemexpress LLC Supplier No. HY1742645MG
DMG-PEG-NHS, MW 2000 is a compound composed of Myristic acid (HY-N2041) and PEG linked by an amide bond, with NHS as an active ester group attached to the end of the PEG chain.
  • Used for protein/biomolecule modification.
  • Supports gene delivery.
  • Applicable in biosensor applications.