Learn More
Medchemexpress LLC DMG-PEG-NHS, MW 2000 | 95.0% | 2000 | 50 MG
SDP
Catalog No. 5000386415
Encompass_Preferred
DMG-PEG-NHS, MW 2000 is a compound composed of Myristic acid and PEG linked by an amide bond, with NHS as an active ester group attached to the end of the PEG chain. It is suitable for various laboratory applications.
  • Used for protein/biomolecule modification
  • Suitable for gene delivery applications
  • Applicable in biosensor development
Catalog No. 50-003-86415 Supplier Medchemexpress LLC Supplier No. HY17426450MG
DMG-PEG-NHS, MW 2000 is a compound composed of Myristic acid and PEG linked by an amide bond, with NHS as an active ester group attached to the end of the PEG chain. It is suitable for various laboratory applications.
  • Used for protein/biomolecule modification
  • Suitable for gene delivery applications
  • Applicable in biosensor development