Learn More
Apexbio Technology LLC DMG-PEG 2000 250mg
SDP
Catalog No. 503107105
Encompass_Preferred
DMG-PEG 2000 can be used in the preparation of liposomes for siRNA delivery to enhance in vitro transfection efficiency It can also be employed in the formulation of lipid nanoparticles (LNP) to improve their mucus penetration and delivery efficiency as well as for oral plasmid DNA delivery methods in vivo
Catalog No. 50-310-7105 Supplier Apexbio Technology LLC Supplier No. M2001250
DMG-PEG 2000 can be used in the preparation of liposomes for siRNA delivery to enhance in vitro transfection efficiency It can also be employed in the formulation of lipid nanoparticles (LNP) to improve their mucus penetration and delivery efficiency as well as for oral plasmid DNA delivery methods in vivo