Learn More
Kyfora Bio DMG-PEG2000 500 MG
Catalog No. 502559805
Encompass_Preferred
DMG-PEG 2000 (1,2-dimyristoyl-rac-glycero-3-methoxypolyethylene glycol-2000) is a PEGylated lipid used in the formation of lipid nanoparticles (LNP) for the development of mRNA vaccines such as COVID-19 vaccine and for the delivery of nucleic acids such as RNAis. It is derived from dimyristoyl glycerol (DMG) which behaves similar to phospholipids, namely in the way they associate with membranes. PEGylation of the lipid enhances bioavailability and prevents opsonization of the LNP. This product is for research use only. Key application(s): mRNA vaccine, gene delivery
Catalog No. 50-255-9805 Supplier Kyfora Bio Supplier No. 14493500
DMG-PEG 2000 (1,2-dimyristoyl-rac-glycero-3-methoxypolyethylene glycol-2000) is a PEGylated lipid used in the formation of lipid nanoparticles (LNP) for the development of mRNA vaccines such as COVID-19 vaccine and for the delivery of nucleic acids such as RNAis. It is derived from dimyristoyl glycerol (DMG) which behaves similar to phospholipids, namely in the way they associate with membranes. PEGylation of the lipid enhances bioavailability and prevents opsonization of the LNP. This product is for research use only. Key application(s): mRNA vaccine, gene delivery