Learn More
Apexbio Technology LLC DMG-PEG2000-Biotin(Synonyms: DMG-PEG-Biotin, Biotinylated DMG-PEG2000, DMG-PEG2000-Biotin Conjugate), 100mg.
SDP
Catalog No. 5000061377
Encompass_Preferred
DMG-PEG2000-Biotin is a synthetic amphiphilic molecule composed of dimyristoyl glycerol (DMG) polyethylene glycol (PEG2000) and biotin functionalities It is commonly employed in biomedical research for surface modification of lipid-based delivery platforms such as lipid nanoparticles (LNPs) and liposomes The PEG moiety enhances colloidal stability prolongs circulation time and reduces rapid clearance in vivo The biotin termination facilitates conjugation to streptavidin-tagged biomolecules exploiting the highly specific biotin-streptavidin interaction Thus DMG-PEG2000-Biotin is valuable for drug-delivery optimization targeted therapeutic approaches and in vitro diagnostic assay development
Catalog No. 50-000-61377 Supplier Apexbio Technology LLC Supplier No. M2007100
DMG-PEG2000-Biotin is a synthetic amphiphilic molecule composed of dimyristoyl glycerol (DMG) polyethylene glycol (PEG2000) and biotin functionalities It is commonly employed in biomedical research for surface modification of lipid-based delivery platforms such as lipid nanoparticles (LNPs) and liposomes The PEG moiety enhances colloidal stability prolongs circulation time and reduces rapid clearance in vivo The biotin termination facilitates conjugation to streptavidin-tagged biomolecules exploiting the highly specific biotin-streptavidin interaction Thus DMG-PEG2000-Biotin is valuable for drug-delivery optimization targeted therapeutic approaches and in vitro diagnostic assay development