Learn More
Apexbio Technology LLC DMG-PEG2000-MAL(Synonyms: DMG-PEG-Maleimide, Maleimide-PEG2000-DMG, DMG-polyethylene glycol2000-Maleimide), 500mg.
SDP
Catalog No. 5000061121
Encompass_Preferred
DMG-PEG2000-MAL is a polyethylene glycol (PEG) derivative functionalized with dimyristoyl glycerol (DMG) and a terminal maleimide (MAL) group The maleimide moiety readily reacts with thiol (-SH) groups present on biomolecules forming stable covalent thioether bonds This reaction facilitates site-specific conjugation or labeling of proteins peptides and other thiol-containing biological structures DMG-PEG2000-MAL is widely utilized in biomedical research particularly in formulating lipid-based drug delivery platforms such as siRNA-loaded liposomes and lipid nanoparticles (LNPs)
Catalog No. 50-000-61121 Supplier Apexbio Technology LLC Supplier No. M2005500
DMG-PEG2000-MAL is a polyethylene glycol (PEG) derivative functionalized with dimyristoyl glycerol (DMG) and a terminal maleimide (MAL) group The maleimide moiety readily reacts with thiol (-SH) groups present on biomolecules forming stable covalent thioether bonds This reaction facilitates site-specific conjugation or labeling of proteins peptides and other thiol-containing biological structures DMG-PEG2000-MAL is widely utilized in biomedical research particularly in formulating lipid-based drug delivery platforms such as siRNA-loaded liposomes and lipid nanoparticles (LNPs)