Learn More
Apexbio Technology LLC DMG-PEG2000-N3 100mg
SDP
Catalog No. 5000097223
Encompass_Preferred
DMG-PEG2000-N3 is a PEGylated lipid derivative utilized for the functionalization of lipid-based drug delivery systems It consists of 1 2-dimyristoyl-sn-glycerol (DMG) polyethylene glycol (PEG 2000 Da) and a terminal azide group (N3) The azide functional group enables selective conjugation via copper-catalyzed azide-alkyne cycloaddition (CuAAC or click chemistry) allowing controlled attachment of biomolecules small-molecule drugs fluorescent probes or targeting ligands onto lipid bilayers DMG-PEG2000-N3 is widely applied in the development of liposomal or nanoparticle formulations for targeted drug delivery and imaging in biomedical research
Catalog No. 50-000-97223 Supplier Apexbio Technology LLC Supplier No. M2012100
DMG-PEG2000-N3 is a PEGylated lipid derivative utilized for the functionalization of lipid-based drug delivery systems It consists of 1 2-dimyristoyl-sn-glycerol (DMG) polyethylene glycol (PEG 2000 Da) and a terminal azide group (N3) The azide functional group enables selective conjugation via copper-catalyzed azide-alkyne cycloaddition (CuAAC or click chemistry) allowing controlled attachment of biomolecules small-molecule drugs fluorescent probes or targeting ligands onto lipid bilayers DMG-PEG2000-N3 is widely applied in the development of liposomal or nanoparticle formulations for targeted drug delivery and imaging in biomedical research