Learn More
Apexbio Technology LLC DMG-PEG2000-NH2-MW-2000-25MG
SDP
Catalog No. 5000567641
Encompass_Preferred
DMG-PEG2000-NH (MW 2000) is a polyethylene glycol (PEG) derivative frequently used in biomedical research as a drug delivery vehicle and stabilizing agent Originating from its ability to enhance the solubility and bioavailability of therapeutic compounds it operates by attaching to drug molecules or nanoparticles forming a protective hydrophilic layer that can improve circulation time and distribution in biological systems The polymer exhibits effective activity in the nanomolar to low micromolar range commonly employed in various in vitro and in vivo studies including cell models and animal research It plays a crucial role in drug screening processes where enhanced delivery and distribution are required Typical applications involve examining its effects within specific cellular pathways and evaluating pharmacokinetic profiles Experimental concentrations typically depend on the specific research design
Catalog No. 50-005-67641 Supplier Apexbio Technology LLC Supplier No. M201525
DMG-PEG2000-NH (MW 2000) is a polyethylene glycol (PEG) derivative frequently used in biomedical research as a drug delivery vehicle and stabilizing agent Originating from its ability to enhance the solubility and bioavailability of therapeutic compounds it operates by attaching to drug molecules or nanoparticles forming a protective hydrophilic layer that can improve circulation time and distribution in biological systems The polymer exhibits effective activity in the nanomolar to low micromolar range commonly employed in various in vitro and in vivo studies including cell models and animal research It plays a crucial role in drug screening processes where enhanced delivery and distribution are required Typical applications involve examining its effects within specific cellular pathways and evaluating pharmacokinetic profiles Experimental concentrations typically depend on the specific research design