Learn More
Apexbio Technology LLC DMG-PEG2000-NHS(Synonyms: DMG-PEG2000-NHS ester, NHS-PEG2000-DMG, DMG PEG NHS ester), 100mg.
SDP
Catalog No. 5000061401
Encompass_Preferred
DMG-PEG2000-NHS is a polyethylene glycol (PEG) derivative functionalized with an N-hydroxysuccinimide (NHS) ester The reactive NHS ester readily forms stable amide linkages upon interaction with primary amine (-NH ) groups facilitating covalent attachment to peptides proteins or amino-functionalized small molecules Due to these conjugation properties DMG-PEG2000-NHS is commonly employed in biomedical research for constructing drug delivery platforms such as lipid nanoparticles (LNPs) or liposomes particularly for siRNA delivery systems
Catalog No. 50-000-61401 Supplier Apexbio Technology LLC Supplier No. M2004100
DMG-PEG2000-NHS is a polyethylene glycol (PEG) derivative functionalized with an N-hydroxysuccinimide (NHS) ester The reactive NHS ester readily forms stable amide linkages upon interaction with primary amine (-NH ) groups facilitating covalent attachment to peptides proteins or amino-functionalized small molecules Due to these conjugation properties DMG-PEG2000-NHS is commonly employed in biomedical research for constructing drug delivery platforms such as lipid nanoparticles (LNPs) or liposomes particularly for siRNA delivery systems