Learn More
Aviva Systems Biology Corp DMGDH Antibody - C-terminal region (ARP64603_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP64603_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DMGDH. It was validated on Western Blot. . Alternate names: DMGDHD, ME2GLYDHApplicationWB. Species reactivity: Horse, Human, Pig, Rat
Catalog No. 50-288-312