Learn More
Medchemexpress LLC HY-12273 100mg , DMH-1 CAS:1206711-16-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY12273100MG
Encompass_Preferred
HY-12273 100mg DMH-1 CAS: 1206711-16-1 DMH-1 is a potent and selective BMP inhibitor with IC50s of 27/107.9/<5/47.6 nM for ALK1/ALK2/ALK3/ALK6, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9655