Learn More
Cayman Chemical DMHAPC-Chol, Purity: ≥95%, CAS: 794494-38-5, 25 mg
SDP
Catalog No. 502933844
Encompass_Preferred
A cationic cholesterol; a component of liposomes used for DNA plasmid delivery in vitro and in vivo in a B16-F10 mouse xenograft model; cytotoxic to B16-F10 cells when used in liposomes at lipid concentrations greater than 20 µM; a component of a DOPE-containing lipocomplex used to deliver DNA into mouse lung via intratracheal injection and to deliver VEGF siRNA into A431 and MDA-MB-231 cells
Catalog No. 50-293-3844 Supplier Cayman Chemical Supplier No. 2658225 MG
A cationic cholesterol; a component of liposomes used for DNA plasmid delivery in vitro and in vivo in a B16-F10 mouse xenograft model; cytotoxic to B16-F10 cells when used in liposomes at lipid concentrations greater than 20 µM; a component of a DOPE-containing lipocomplex used to deliver DNA into mouse lung via intratracheal injection and to deliver VEGF siRNA into A431 and MDA-MB-231 cells