Learn More
Cayman Chemical DMHAPC-Chol 5mg
SDP
Catalog No. 502933665
Encompass_Preferred
A cationic cholesterol; a component of liposomes used for DNA plasmid delivery in vitro and in vivo in a B16-F10 mouse xenograft model; cytotoxic to B16-F10 cells when used in liposomes at lipid concentrations greater than 20 µM; a component of a DOPE-containing lipocomplex used to deliver DNA into mouse lung via intratracheal injection and to deliver VEGF siRNA into A431 and MDA-MB-231 cells
Catalog No. 50-293-3665 Supplier Cayman Chemical Supplier No. 265825 MG
A cationic cholesterol; a component of liposomes used for DNA plasmid delivery in vitro and in vivo in a B16-F10 mouse xenograft model; cytotoxic to B16-F10 cells when used in liposomes at lipid concentrations greater than 20 µM; a component of a DOPE-containing lipocomplex used to deliver DNA into mouse lung via intratracheal injection and to deliver VEGF siRNA into A431 and MDA-MB-231 cells