Learn More
Medchemexpress LLC DMHCA | 79066-03-8 | 99.9% | 50 MG
SDP
Catalog No. 5000340613
Encompass_Preferred
DMHCA is a potent and selective LXR agonist that specifically activates the cholesterol efflux arm of the LXR pathway without stimulating triglyceride synthesis. It also exhibits anti-inflammatory effects and is suitable for research on cholesterol homeostasis and diabetes.
  • Potent and selective LXR agonist.
  • Activates cholesterol efflux pathway.
  • Exhibits anti-inflammatory effects.
  • Suitable for research on cholesterol homeostasis.
  • Suitable for research on diabetes.
  • Restores membrane fluidity in diabetic cells ex vivo.
  • Enhances erythrocyte progenitor production.
  • Restores cholesterol homeostasis in diabetic retina in vivo.
Catalog No. 50-003-40613 Supplier Medchemexpress LLC Supplier No. HY12909850MG
DMHCA is a potent and selective LXR agonist that specifically activates the cholesterol efflux arm of the LXR pathway without stimulating triglyceride synthesis. It also exhibits anti-inflammatory effects and is suitable for research on cholesterol homeostasis and diabetes.
  • Potent and selective LXR agonist.
  • Activates cholesterol efflux pathway.
  • Exhibits anti-inflammatory effects.
  • Suitable for research on cholesterol homeostasis.
  • Suitable for research on diabetes.
  • Restores membrane fluidity in diabetic cells ex vivo.
  • Enhances erythrocyte progenitor production.
  • Restores cholesterol homeostasis in diabetic retina in vivo.