Learn More
Medchemexpress LLC Dmhca | 79066-03-8 | MFCD12404996 | ≥98.0% | 401.63 g/mol | C26H43NO2 | 5 MG
SDP
Catalog No. 5000263268
Encompass_Preferred
DMHCA is a selective Liver X Receptor (LXR) agonist used as a research reagent to activate the cholesterol efflux arm of LXR signaling while minimizing induction of triglyceride synthesis. It is commonly used in studies of cholesterol metabolism, lipid homeostasis, and inflammation.
  • Selective LXR agonist that promotes cholesterol efflux.
  • Minimal stimulation of triglyceride synthesis compared with other LXR agonists.
  • Applicable to studies of cholesterol metabolism, lipid homeostasis, and inflammatory responses.
  • Characterized by CAS 79066-03-8, molecular formula C26H43NO2, molecular weight 401.63 g/mol.
  • Supplied in small research pack sizes suitable for in vitro studies.
  • Reported purity ≥98% by HPLC from third-party suppliers.
Catalog No. 50-002-63268 Supplier Medchemexpress LLC Supplier No. HY1290985MG
DMHCA is a selective Liver X Receptor (LXR) agonist used as a research reagent to activate the cholesterol efflux arm of LXR signaling while minimizing induction of triglyceride synthesis. It is commonly used in studies of cholesterol metabolism, lipid homeostasis, and inflammation.
  • Selective LXR agonist that promotes cholesterol efflux.
  • Minimal stimulation of triglyceride synthesis compared with other LXR agonists.
  • Applicable to studies of cholesterol metabolism, lipid homeostasis, and inflammatory responses.
  • Characterized by CAS 79066-03-8, molecular formula C26H43NO2, molecular weight 401.63 g/mol.
  • Supplied in small research pack sizes suitable for in vitro studies.
  • Reported purity ≥98% by HPLC from third-party suppliers.