Learn More
Cole-Parmer Fluke 114 Electrical True RMS Multimeter
Catalog No. 50992550 Shop All Cole-Parmer Products
Encompass
The Fluke 114 is the troubleshooting tool for ''go/no-go'' testing. It includes a feature to prevent false readings caused by ghost voltage. General specifications include: accuracy specified for 1 year after calibration, at operating temperatures of 18°C, with relative humidity at 0% to 90%. Available for sale in the United States and Canada only.
Catalog No. 50-992-550 Supplier Cole-Parmer Supplier No. 2000559
Add to Cart
Add to Cart
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
The Fluke 114 is the troubleshooting tool for ''go/no-go'' testing. It includes a feature to prevent false readings caused by ghost voltage. General specifications include: accuracy specified for 1 year after calibration, at operating temperatures of 18°C, with relative humidity at 0% to 90%. Available for sale in the United States and Canada only.