Learn More
Cole-Parmer Fluke 117 Electrician's Multimeter with True-RMS
Supplier:  Cole-Parmer 2601632
Encompass
These compact meters feature True-RMS AC and DC voltage, 6000-count resolution, min/max/average capabilities, frequency, and capacitance. Units include a large display and white LED backlight for working in poorly lit environments. Fluke Model 117 is ideal for electricians and use in demanding conditions. Unit includes VoltAlert™ technology for integrated noncontact voltage selection, AutoVolt feature provides automatic AC/DC voltage selection. Low input impedance prevents false readings due to ghost voltage. Available for sale in the United States and Canada only.
Catalog No. 50-992-552