Learn More
Cole-Parmer Amprobe DM73C Meterman Probe Style Pocket Digital Multimeter
Catalog No. 50993178 Shop All Cole-Parmer Products
Encompass
DM78B Credit Card Sized DMM
  • Autoranging
  • UL1244 / IEC1010-1 Approved
  • Pocket sized meter
  • The resolution and measuring functions of larger, more expensive meters
  • Analog Bargraph
  • Datahold
  • Auto Power Off
  • Shipped with test leads and probes attached, batteries installed, its own protective vinyl carry case and manual
    DM73C - Smallest Probe Style DMM
  • Easy, one-hand operation
  • CAT II 600V, CAT III 300V Rated, UL3111-1, CE
  • Superior 0.5% Accuracy
  • Auto Power Off
  • Analog Bargraph
  • Data Hold
  • Shipped with spare probe tips, alligator clip for ground test lead, two batteries installed
  • Catalog No. 50-993-178 Supplier Cole-Parmer Supplier No. 2000555
    DM78B Credit Card Sized DMM
  • Autoranging
  • UL1244 / IEC1010-1 Approved
  • Pocket sized meter
  • The resolution and measuring functions of larger, more expensive meters
  • Analog Bargraph
  • Datahold
  • Auto Power Off
  • Shipped with test leads and probes attached, batteries installed, its own protective vinyl carry case and manual
    DM73C - Smallest Probe Style DMM
  • Easy, one-hand operation
  • CAT II 600V, CAT III 300V Rated, UL3111-1, CE
  • Superior 0.5% Accuracy
  • Auto Power Off
  • Analog Bargraph
  • Data Hold
  • Shipped with spare probe tips, alligator clip for ground test lead, two batteries installed
  • Product Title