Learn More
Cole-Parmer Amprobe 37XR DMM, T-RMS with Comp Logic Test
Supplier:  Cole-Parmer 2000550
Encompass
True RMS with component and logic test, its inductance is up to 40 H; frequency to 40 MHz, capacitance to 400 F, and logic to 20 MHz. It also has an input warning tone when test leads are in wrong jack; 10,000 count; 0.1% accuracy; 1000 V/750 V dc/ac True RMS, and 10 Amps ac/dc, fully safety fused.
Catalog No. 50-993-175