Learn More
Cole-Parmer Fluke 289 True RMS Industrial Logging Multimeter with TrendCapture
Catalog No. 50993317 Shop All Cole-Parmer Products
Encompass
This industrial meter for demanding users features built-in logger records reading and time of day to catch intermittent problems. The data can be viewed on the meter in what's called, TrendCapture, or optional FlukeView software. The meter also features peak capture to measure transients as fast as 250 S.
Catalog No. 50-993-317 Supplier Cole-Parmer Supplier No. 2604896
This industrial meter for demanding users features built-in logger records reading and time of day to catch intermittent problems. The data can be viewed on the meter in what's called, TrendCapture, or optional FlukeView software. The meter also features peak capture to measure transients as fast as 250 S.