Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC DMNB | 20357-25-9 | 100.0% | 211.17 | C9H9NO5 | 25 G
SDP

Catalog No. 5000214511
Encompass_Preferred

DMNB (6-nitroveratraldehyde) is a high-purity chemical intermediate used as a precursor in radiochemical synthesis, notably for preparing no-carrier-added 6-[18F]fluoro-L-DOPA for PET studies. The solid reagent is provided with analytical documentation and recommended storage to preserve stability for research applications.

  • High purity: 99.98% (HPLC).
  • Chemical formula C9H9NO5 and molecular weight 211.17.
  • Used as a precursor for synthesis of no-carrier-added 6-[18F]fluoro-L-DOPA.
  • Storage: store at 4°C under inert atmosphere; in solvent: -80°C (6 months), -20°C (1 month).
  • Supplied with a certificate of analysis and safety data sheet.

Catalog No. 50-002-14511 Supplier Medchemexpress LLC Supplier No. HY10070525G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

DMNB (6-nitroveratraldehyde) is a high-purity chemical intermediate used as a precursor in radiochemical synthesis, notably for preparing no-carrier-added 6-[18F]fluoro-L-DOPA for PET studies. The solid reagent is provided with analytical documentation and recommended storage to preserve stability for research applications.

  • High purity: 99.98% (HPLC).
  • Chemical formula C9H9NO5 and molecular weight 211.17.
  • Used as a precursor for synthesis of no-carrier-added 6-[18F]fluoro-L-DOPA.
  • Storage: store at 4°C under inert atmosphere; in solvent: -80°C (6 months), -20°C (1 month).
  • Supplied with a certificate of analysis and safety data sheet.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.