Learn More
Cayman Chemical D-Mannose-6-Phosphate (sodium salt hydrate), Purity: ≥95%, 100 mg
SDP
Catalog No. 502982853
Encompass_Preferred
A phosphorylated aldohexose that can be used to produce glucose, mannitol, or N-glycans; also a ligand for both cation-dependent and -independent receptors and binds, as part of an N-glycan, to MRH domains on other proteins
Catalog No. 50-298-2853 Supplier Cayman Chemical Supplier No. 18890100 MG
A phosphorylated aldohexose that can be used to produce glucose, mannitol, or N-glycans; also a ligand for both cation-dependent and -independent receptors and binds, as part of an N-glycan, to MRH domains on other proteins