Learn More
Biotium DMNP-EDTA (Caged Calcium)
Catalog No. 501964701
Encompass_Preferred
DMNP-EDTA (also known as DM-Nitrophen)  is a caged Ca2 chelator. Upon photolysis, the Kd for Ca2 increases from 5 nM to 3 mM, resulting in a pulse of free Ca2. Properties: Light yellow solid soluble in water (pH >6) and DMSO; Store desiccated at -20°C and protect from light. Aqueous solutions stable for at least three months when stored at 4°C, protected from light ; C18H19K4N3O12 MW: 626.
Catalog No. 50-196-4701 Supplier Biotium Supplier No. 50050
DMNP-EDTA (also known as DM-Nitrophen)  is a caged Ca2 chelator. Upon photolysis, the Kd for Ca2 increases from 5 nM to 3 mM, resulting in a pulse of free Ca2. Properties: Light yellow solid soluble in water (pH >6) and DMSO; Store desiccated at -20°C and protect from light. Aqueous solutions stable for at least three months when stored at 4°C, protected from light ; C18H19K4N3O12 MW: 626.