Learn More
Selleck Chemical LLC DMNQ
SDP
Catalog No. 5000002926
Encompass_Preferred
DMNQ is a 1 4-naphthoquinone that acts as a redox-cycling agent typically increasing intracellular superoxide and hydrogen peroxide formation
Catalog No. 50-000-02926 Supplier Selleck Chemical LLC Supplier No. S65632MG
DMNQ is a 1 4-naphthoquinone that acts as a redox-cycling agent typically increasing intracellular superoxide and hydrogen peroxide formation